3-Fluoro-2,6-dimethoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

55585-500MG 55585-1G 55585-5G
77339-034EA 545.9 USD
77339-034 77353-366 77377-640
3-Fluoro-2,6-dimethoxybenzoic acid ≥95%
3-Fluoro-2,6-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₄
Storage Temperature: Ambient
MDL Number: MFCD24480409
CAS Number: 52189-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR