3-(2-Bromo-5-methoxyphenyl)-3-hydroxypropanoic acid ≥97%

Supplier: AOBChem USA

Warning

55570-500MG 55570-1G 55570-5G
77339-606EA 603.58 USD
77339-606 77353-364 77377-634
3-(2-Bromo-5-methoxyphenyl)-3-hydroxypropanoic acid ≥97%
3-(2-Bromo-5-methoxyphenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₄
MW: 275.096 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757541
CAS Number: 1698402-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR