3-(5-(Benzyloxy)-2-bromophenyl)-3-hydroxypropanoic acid ≥97%

Supplier: AOBChem USA

Warning

55568-500MG 55568-1G
77340-472EA 1203.04 USD
77340-472 77353-362
3-(5-(Benzyloxy)-2-bromophenyl)-3-hydroxypropanoic acid ≥97%
3-(5-(Benzyloxy)-2-bromophenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₅BrO₄
MW: 351.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757540
CAS Number: 2901065-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR