Ethyl 3-(5-(benzyloxy)-2-bromophenyl)-3-hydroxypropanoate ≥97%

Supplier: AOBChem USA

Warning

55567-500MG 55567-1G 55567-5G
77340-284EA 879.62 USD
77340-284 77353-360 77377-632
Ethyl 3-(5-(benzyloxy)-2-bromophenyl)-3-hydroxypropanoate ≥97%
Ethyl 3-(5-(benzyloxy)-2-bromophenyl)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₈H₁₉BrO₄
MW: 379.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564309
CAS Number: 2734775-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR