3-(4-Fluorophenyl)-3-hydroxypropanoic acid ≥95%

Supplier: AOBChem USA

Warning

55564-500MG 55564-1G 55564-5G
77329-486EA 318.27 USD
77329-486 77353-358 77377-628
3-(4-Fluorophenyl)-3-hydroxypropanoic acid ≥95%
3-(4-Fluorophenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD16302337
CAS Number: 40620-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR