Ethyl 3-(4-chloro-3-fluorophenyl)-3-hydroxypropanoate ≥97%

Supplier: AOBChem USA

Warning

55557-1G 55557-5G 55557-25G
77341-112EA 71.07 USD
77341-112 77365-782 77377-622
Ethyl 3-(4-chloro-3-fluorophenyl)-3-hydroxypropanoate ≥97%
Ethyl 3-(4-chloro-3-fluorophenyl)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClFO₃
MW: 246.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16756888
CAS Number: 1248697-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR