3-(2,5-Difluorophenyl)-3-hydroxypropanoic acid ≥97%

Supplier: AOBChem USA

Warning

55552-500MG 55552-1G 55552-5G
77339-604EA 603.58 USD
77339-604 77353-352 77377-620
3-(2,5-Difluorophenyl)-3-hydroxypropanoic acid ≥97%
3-(2,5-Difluorophenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16749158
CAS Number: 1249279-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR