1-(2-Chloro-3-methoxypyridin-4-yl)ethanol ≥97%

Supplier: AOBChem USA

Warning

55530-500MG 55530-1G 55530-5G 55530-25G
77329-478EA 120.51 USD
77329-478 77343-750 77365-774 77377-616
1-(2-Chloro-3-methoxypyridin-4-yl)ethanol ≥97%
1-(2-Chloro-3-methoxypyridin-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₂
MW: 187.624 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564308
CAS Number: 2734779-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR