Ethyl 6-fluoro-3-iodo-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

55529-500MG 55529-1G 55529-5G
77329-476EA 309 USD
77329-476 77353-348 77377-614
Ethyl 6-fluoro-3-iodo-2-methoxybenzoate ≥97%
Ethyl 6-fluoro-3-iodo-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀FIO₃
MW: 324.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757535
CAS Number: 2386006-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR