Methyl 6-fluoro-3-iodo-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

55528-500MG 55528-1G 55528-25G 55528-5G
77329-474EA 292.52 USD
77329-474 77353-346 77387-656 77377-612
Methyl 6-fluoro-3-iodo-2-methoxybenzoate ≥97%
Methyl 6-fluoro-3-iodo-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈FIO₃
Storage Temperature: Ambient
MDL Number: MFCD34757534
CAS Number: 2092791-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR