6-Fluoro-3-iodo-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

55527-500MG 55527-1G 55527-5G 55527-25G
77329-472EA 224.54 USD
77329-472 77353-344 77365-772 77387-654
6-Fluoro-3-iodo-2-methoxybenzoic acid ≥97%
6-Fluoro-3-iodo-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆FIO₃
Storage Temperature: Ambient
MDL Number: MFCD34564307
CAS Number: 2091622-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR