Ethyl 4-bromo-2-chloro-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

55525-500MG 55525-1G 55525-5G
77329-470EA 328.57 USD
77329-470 77353-342 77377-610
Ethyl 4-bromo-2-chloro-5-(trifluoromethyl)benzoate ≥97%
Ethyl 4-bromo-2-chloro-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₇BrClF₃O₂
MW: 331.514 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564306
CAS Number: 2705275-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR