4-Bromo-2-(2-methylbenzyl)phenol ≥97%

Supplier: AOBChem USA

Warning

55521-500MG 55521-1G 55521-5G
77338-842EA 524.27 USD
77338-842 77353-340 77377-602
4-Bromo-2-(2-methylbenzyl)phenol ≥97%
4-Bromo-2-(2-methylbenzyl)phenol

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD34530422
CAS Number: 2734777-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR