2,2-Dichloro-1-(4-methoxy-3-methylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

55502-500MG 55502-1G 55502-5G 55502-25G
77329-462EA 185.4 USD
77329-462 77347-358 77365-764 77387-652
2,2-Dichloro-1-(4-methoxy-3-methylphenyl)ethanol ≥95%
2,2-Dichloro-1-(4-methoxy-3-methylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂Cl₂O₂
MW: 235.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530418
CAS Number: 2734778-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR