5-(3,4-Dichlorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55501-500MG 55501-1G 55501-25G 55501-5G
77329-460EA 297.67 USD
77329-460 77353-338 77387-650 77377-600
5-(3,4-Dichlorophenyl)thiazol-2-amine ≥97%
5-(3,4-Dichlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆Cl₂N₂S
MW: 245.128 g/mol
Storage Temperature: Ambient
CAS Number: 926223-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR