(6-Fluoro-3-iodo-2-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

55497-500MG 55497-1G 55497-5G
77329-456EA 334.75 USD
77329-456 77353-334 77377-598
(6-Fluoro-3-iodo-2-methoxyphenyl)methanol ≥97%
(6-Fluoro-3-iodo-2-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈FIO₂
MW: 282.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757524
CAS Number: 2090747-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR