1-(3,5-Difluoro-4-methylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

55485-500MG 55485-1G 55485-5G 55485-25G
77329-450EA 134.93 USD
77329-450 77344-470 77365-756 77387-644
1-(3,5-Difluoro-4-methylphenyl)ethanol ≥97%
1-(3,5-Difluoro-4-methylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O
MW: 172.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28519443
CAS Number: 1781452-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR