3,5-Difluoro-4-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

55484-500MG 55484-1G 55484-5G 55484-25G
77329-448EA 187.46 USD
77329-448 77353-330 77365-754 77387-642
3,5-Difluoro-4-methyl-1,1'-biphenyl ≥95%
3,5-Difluoro-4-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂
MW: 204.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530414
CAS Number: 2734773-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR