5-Cyclopropyl-1,3-difluoro-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

55483-500MG 55483-1G 55483-5G 55483-25G
77329-446EA 187.46 USD
77329-446 77353-328 77365-752 77387-640
5-Cyclopropyl-1,3-difluoro-2-methylbenzene ≥95%
5-Cyclopropyl-1,3-difluoro-2-methylbenzene
Formula: C₁₀H₁₀F₂
Storage Temperature: Ambient
MDL Number: MFCD34530413
CAS Number: 2734778-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR