3-Chloro-5-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

55482-500MG 55482-1G 55482-5G 55482-25G
77329-444EA 122.57 USD
77329-444 77343-938 77365-750 77377-594
3-Chloro-5-fluoro-1,1'-biphenyl ≥97%
3-Chloro-5-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈ClF
Storage Temperature: Ambient
MDL Number: MFCD34530412
CAS Number: 1651205-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR