Ethyl 6-chloro-2-fluoro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

55460-1G 55460-5G 55460-25G
77341-464EA 88.58 USD
77341-464 77365-746 77377-590
Ethyl 6-chloro-2-fluoro-3-methoxybenzoate ≥97%
Ethyl 6-chloro-2-fluoro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD10566184
CAS Number: 1397181-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR