(6-Chloro-2-fluoro-3-methoxyphenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

55455-1G 55455-5G 55455-25G
77341-792EA 104.03 USD
77341-792 77365-744 77377-588
(6-Chloro-2-fluoro-3-methoxyphenyl)(thiomorpholino)methanone ≥97%
(6-Chloro-2-fluoro-3-methoxyphenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClFNO₂S
MW: 289.754 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530409
CAS Number: 2734777-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR