(6-Chloro-2-fluoro-3-methoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

55453-1G 55453-5G 55453-25G
77341-788EA 104.03 USD
77341-788 77365-740 77377-584
(6-Chloro-2-fluoro-3-methoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(6-Chloro-2-fluoro-3-methoxyphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆ClFN₂O₂
MW: 286.73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530407
CAS Number: 2734778-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR