(6-Chloro-2-fluoro-3-methoxyphenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

55452-1G 55452-5G 55452-25G
77341-786EA 104.03 USD
77341-786 77365-738 77377-582
(6-Chloro-2-fluoro-3-methoxyphenyl)(piperidin-1-yl)methanone ≥97%
(6-Chloro-2-fluoro-3-methoxyphenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₅ClFNO₂
MW: 271.715 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530406
CAS Number: 2734775-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR