(6-Chloro-2-fluoro-3-methoxyphenyl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

55451-1G 55451-5G 55451-25G
77341-784EA 104.03 USD
77341-784 77365-736 77377-580
(6-Chloro-2-fluoro-3-methoxyphenyl)(pyrrolidin-1-yl)methanone ≥97%
(6-Chloro-2-fluoro-3-methoxyphenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClFNO₂
MW: 257.689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530405
CAS Number: 2734776-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR