6-Chloro-2-fluoro-N-isopropyl-3-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

55448-1G 55448-5G 55448-25G
77341-782EA 104.03 USD
77341-782 77365-730 77377-574
6-Chloro-2-fluoro-N-isopropyl-3-methoxybenzamide ≥97%
6-Chloro-2-fluoro-N-isopropyl-3-methoxybenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClFNO₂
MW: 245.678 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530402
CAS Number: 2734777-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR