6-Chloro-2-fluoro-3-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

55447-500MG 55447-1G 55447-5G 55447-25G
77329-440EA 187.46 USD
77329-440 77353-326 77365-728 77387-638
6-Chloro-2-fluoro-3-methoxy-N-methylbenzamide ≥97%
6-Chloro-2-fluoro-3-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉ClFNO₂
MW: 217.625 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530401
CAS Number: 2734776-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR