5-(3-Fluoro-4-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55422-500MG 55422-1G 55422-5G
77329-436EA 350.2 USD
77329-436 77353-322 77377-572
5-(3-Fluoro-4-methylphenyl)thiazol-2-amine ≥97%
5-(3-Fluoro-4-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂S
MW: 208.255 g/mol
Storage Temperature: Ambient
CAS Number: 1552256-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR