2,2-Dichloro-1-(3-chloro-4-methylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

55419-500MG 55419-1G 55419-5G 55419-25G
77329-434EA 88.58 USD
77329-434 77342-326 77365-724 77377-570
2,2-Dichloro-1-(3-chloro-4-methylphenyl)ethanol ≥95%
2,2-Dichloro-1-(3-chloro-4-methylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₉Cl₃O
MW: 239.526 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530398
CAS Number: 2734777-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR