Ethyl 2-(1-hydroxycyclopentyl)acetate ≥97%

Supplier: AOBChem USA

Warning

55396-500MG 55396-1G 55396-5G 55396-25G
77329-432EA 233.81 USD
77329-432 77353-320 77365-720 77387-634
Ethyl 2-(1-hydroxycyclopentyl)acetate ≥97%
Ethyl 2-(1-hydroxycyclopentyl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD01822296
CAS Number: 3197-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR