5-(3,4-Dimethylphenyl)-N-methylthiazol-2-amine ≥95%
Supplier: AOBChem USA
|
Warning
|
55380-500MG
55380-1G
55380-5G
55380-25G
77329-428EA
202.91
USD
77329-428
77353-318
77365-714
77387-632
5-(3,4-Dimethylphenyl)-N-methylthiazol-2-amine ≥95%
5-(3,4-Dimethylphenyl)-N-methylthiazol-2-amine
Precaución: for Research Use Only
Formula:
C₁₂H₁₄N₂S MW: 218.318 g/mol Storage Temperature: Ambient |
CAS Number:
1597251-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...