5-Bromo-2-iodo-1,3-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

55367-1G 55367-5G 55367-25G
77340-738EA 60.77 USD
77340-738 77365-712 77377-562
5-Bromo-2-iodo-1,3-dimethoxybenzene ≥95%
5-Bromo-2-iodo-1,3-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrIO₂
MW: 342.956 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738448
CAS Number: 1037082-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR