2,2-Dichloro-1-(2-methoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

55318-500MG 55318-1G 55318-5G
77329-426EA 318.27 USD
77329-426 77353-316 77377-560
2,2-Dichloro-1-(2-methoxyphenyl)ethanol ≥95%
2,2-Dichloro-1-(2-methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₀Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34530392
CAS Number: 2734778-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR