2-Ethoxy-5-fluoro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

55293-500MG 55293-1G 55293-5G 55293-25G
77329-424EA 189.52 USD
77329-424 77353-314 77365-710 77387-630
2-Ethoxy-5-fluoro-4-iodopyridine ≥97%
2-Ethoxy-5-fluoro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₇H₇FINO
Storage Temperature: Ambient
MDL Number: MFCD28795533
CAS Number: 1803782-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR