5-(3,5-Dichlorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55288-500MG 55288-1G 55288-5G
77329-420EA 347.11 USD
77329-420 77353-310 77377-558
5-(3,5-Dichlorophenyl)thiazol-2-amine ≥97%
5-(3,5-Dichlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆Cl₂N₂S
MW: 245.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564296
CAS Number: 1603310-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR