3-Amino-4,6-dibromo-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

55284-500MG 55284-1G 55284-5G
77329-418EA 324.45 USD
77329-418 77353-308 77377-556
3-Amino-4,6-dibromo-2-methylbenzoic acid ≥97%
3-Amino-4,6-dibromo-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇Br₂NO₂
MW: 308.955 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530388
CAS Number: 1935932-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR