5-(4-Bromophenyl)-N-methylthiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55266-500MG 55266-1G 55266-5G 55266-25G
77329-416EA 266.77 USD
77329-416 77353-302 77365-706 77387-626
5-(4-Bromophenyl)-N-methylthiazol-2-amine ≥97%
5-(4-Bromophenyl)-N-methylthiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂S
MW: 269.161 g/mol
Storage Temperature: Ambient
CAS Number: 1592334-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR