5-Mesitylthiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55258-500MG 55258-1G 55258-5G 55258-25G
77329-414EA 269.86 USD
77329-414 77353-300 77365-704 77387-624
5-Mesitylthiazol-2-amine ≥97%
5-Mesitylthiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂S
Storage Temperature: Ambient
CAS Number: 1179313-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR