5-([1,1'-Biphenyl]-4-yl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55256-500MG 55256-1G 55256-5G 55256-25G
77329-412EA 182.31 USD
77329-412 77346-968 77365-702 77387-622
5-([1,1'-Biphenyl]-4-yl)thiazol-2-amine ≥97%
5-([1,1'-Biphenyl]-4-yl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂S
MW: 252.334 g/mol
Storage Temperature: Ambient
CAS Number: 1178275-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR