5-Bromo-2-fluoro-3,3'-dimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

55245-500MG 55245-1G 55245-5G
77338-828EA 523.24 USD
77338-828 77353-292 77377-544
5-Bromo-2-fluoro-3,3'-dimethyl-1,1'-biphenyl ≥95%
5-Bromo-2-fluoro-3,3'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrF
MW: 279.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757469
CAS Number: 2807446-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR