5-Bromo-2-fluoro-3,4'-dimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

55244-500MG 55244-1G 55244-5G
77339-820EA 651.99 USD
77339-820 77353-290 77377-542
5-Bromo-2-fluoro-3,4'-dimethyl-1,1'-biphenyl ≥95%
5-Bromo-2-fluoro-3,4'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrF
MW: 279.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757468
CAS Number: 2807440-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR