5-Bromo-2-fluoro-2',3-dimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

55243-500MG 55243-1G 55243-5G
77338-826EA 523.24 USD
77338-826 77353-288 77377-540
5-Bromo-2-fluoro-2',3-dimethyl-1,1'-biphenyl ≥95%
5-Bromo-2-fluoro-2',3-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrF
MW: 279.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757467
CAS Number: 2807441-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR