5-(Benzyloxy)-2,4-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

55231-500MG 55231-1G
77340-440EA 1075.32 USD
77340-440 77353-282
5-(Benzyloxy)-2,4-dimethylbenzaldehyde ≥97%
5-(Benzyloxy)-2,4-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₂
MW: 240.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757459
CAS Number: 2901106-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR