5-(Benzyloxy)-2,4-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

55229-500MG 55229-1G 55229-5G
77340-176EA 816.79 USD
77340-176 77353-280 77377-534
5-(Benzyloxy)-2,4-dimethylbenzoic acid ≥97%
5-(Benzyloxy)-2,4-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MW: 256.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757457
CAS Number: 2901102-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR