2,2-Dichloro-1-(4-chloro-2-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

55206-500MG 55206-1G 55206-5G 55206-25G
77329-410EA 185.4 USD
77329-410 77347-356 77365-700 77387-620
2,2-Dichloro-1-(4-chloro-2-fluorophenyl)ethanol ≥95%
2,2-Dichloro-1-(4-chloro-2-fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₆Cl₃FO
Storage Temperature: Ambient
MDL Number: MFCD34530370
CAS Number: 2734773-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR