2-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

55201-500MG 55201-1G 55201-5G 55201-25G
77329-406EA 97.85 USD
77329-406 77342-716 77365-696 77377-530
2-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid ≥95%
2-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃F₄NO₄
Storage Temperature: Ambient
MDL Number: MFCD22273734
CAS Number: 1389313-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR