5-(2,5-Dimethylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55129-500MG 55129-1G 55129-5G
77338-960EA 536.63 USD
77338-960 77353-274 77377-528
5-(2,5-Dimethylphenyl)thiazol-2-amine ≥97%
5-(2,5-Dimethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂S
MW: 204.291 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00629043
CAS Number: 303099-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR