Ethyl 5-chloro-2-fluoro-4-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

55119-500MG 55119-1G 55119-5G 55119-25G
77329-402EA 281.19 USD
77329-402 77353-272 77365-692 77387-616
Ethyl 5-chloro-2-fluoro-4-(trifluoromethyl)benzoate ≥97%
Ethyl 5-chloro-2-fluoro-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₇ClF₄O₂
MW: 270.608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752619
CAS Number: 1805109-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR