Methyl 5-chloro-2-fluoro-4-(trifluoromethyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

55118-500MG 55118-1G 55118-5G
77329-400EA 334.75 USD
77329-400 77353-270 77377-524
Methyl 5-chloro-2-fluoro-4-(trifluoromethyl)benzoate ≥95%
Methyl 5-chloro-2-fluoro-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₅ClF₄O₂
MW: 256.58141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752646
CAS Number: 1805524-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR