5-Chloro-2-fluoro-4-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

55116-500MG 55116-1G 55116-5G
77335-562EA 357.41 USD
77335-562 77353-268 77377-522
5-Chloro-2-fluoro-4-(trifluoromethyl)phenol ≥97%
5-Chloro-2-fluoro-4-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₃ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD34757442
CAS Number: 116640-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR